3,4-Dinitrobenzonitrile 97%

Danger

A13131-06 A13131-03 A13131-03 A13131-06
AAA13131-03EA 828.38 USD
CAAAA13131-03 AAA13131-03 AAA13131-06 CAAAA13131-06
3,4-Dinitrobenzonitrile 97%
3,4-Dinitrobenzonitrile
Formula: C₇H₃N₃O₄
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00013370
CAS Number: 4248-33-3
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR