Cyanoacetamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Cyanoacetamide
A12482-22
A12482-22
A12482-36
A12482-0E
A12482-0E
A12482-36
AAA12482-22EA
32.49
USD
AAA12482-22
CAAAA12482-36
CAAAA12482-22
AAA12482-36
AAAA12482-0E
CAAAAA12482-0E
Cyanoacetamide 99%
Cyanoacetamide
Formula:
C₃H₄N₂O MW: 84.08 g/mol Melting Pt: 118…122 °C Flash Pt: 215 °C(419 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008024 CAS Number: 107-91-5 EINECS: 203-531-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...