DL-Citrulline 97%

A11386-14 A11386-06 A11386-22
AAA11386-06EA 49.4 USD
AAA11386-06 AAA11386-14 AAA11386-22
DL-Citrulline 97%
DL-Citrulline
Formula: C₆H₁₃N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00007955
CAS Number: 627-77-0
EINECS: 211-012-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR