5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 99%

Warning

Synonyms: 5,5-Dimethyloxazolidine-2,4-dione

A10714-14 A10714-06 A10714-03
CAAAA10714-06 CAAAA10714-14 CAAAA10714-03
5,5-Dimethyloxazolidine-2,4-dione (dimethadione) 99%
5,5-Dimethyloxazolidine-2,4-dione (Dimethadione)
Formula: C₅H₇NO₃
MW: 129.12 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005379
CAS Number: 695-53-4
EINECS: 211-781-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR