Benzaldehyde 99+%

Supplier: Thermo Scientific Chemicals
Warning

A10348-0B A10348-30 A10348-30 A10348-0I
AAA10348-0BEA 72 USD
AAA10348-0B AAA10348-0I AAA10348-30 CAAAA10348-30
Benzaldehyde 99+%
Benzaldehyde
Formula: C₆H₅CHO
MW: 106.12 g/mol
Boiling Pt: 178…179 °C
Melting Pt: –26 °C
Density: 1.047
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: "9",III
Merck Index: 13,01057
https://us.vwr.com/stibo/web/std.lang.all/60/95/746095.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR