2-Nitrocinnamaldehyde predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2'-Nitrocinnamaldehyde

A10308-18 A10308-18 A10308-09 A10308-14 A10308-14 A10308-09
AAA10308-18EA 181.99 USD
AAA10308-18 AAAA10308-14 AAA10308-09 CAAAA10308-09 CAAAA10308-18 CAAAAA10308-14
2-Nitrocinnamaldehyde predominantly trans 98%
2-Nitrocinnamaldehyde
Formula: C₉H₇NO₃
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00007188
CAS Number: 1466-88-2
EINECS: 215-988-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR