Lanthanum, sputtering target, Diameter 50.8 mm, Thickness 1.59 mm

Danger

45740-KS
AA45740-KSEA 897.99 USD
AA45740-KS
Lanthanum, sputtering target, Diameter 50.8 mm, Thickness 1.59 mm
Lanthanum
Formula: La
MW: 138.91 g/mol
Boiling Pt: 3464 °C (1013 hPa)
Melting Pt: 920 °C
Density: ∼6.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
UN: 3208
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR