Dysprosium, sputtering target, Diameter 50.8 mm, Thickness 1.59 mm

Supplier: Thermo Scientific Chemicals

045735-KS
AA45735-KSEA 2098.99 USD
AA45735-KS
Dysprosium, sputtering target, Diameter 50.8 mm, Thickness 1.59 mm
Dysprosium
Formula: Dy
MW: 162.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR