Calcium thioglycolate trihydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium thioglycolate trihydrateMercaptoacetic acid calcium salt trihydrateCalcium bis(mercaptoacetate) trihydrateCalcium mercaptoacetate trihydrate

45557-18
AA45557-18EA 110.99 USD
AA45557-18
Calcium thioglycolate trihydrate 98%
Calcium bis(mercaptoacetate) (Calcium thioglycolate) trihydrate
Formula: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
Melting Pt: 280…290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150707
CAS Number: 65208-41-5
EINECS: 212-402-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR