Iridium ≥99.8% (metals basis), slug, Length 12.0 mm (0.48 in), Diameter 6.0 mm (0.24 in)

045121-KG
AA45121-KGEA 4673.99 USD
AA45121-KG
Iridium ≥99.8% (metals basis), slug, Length 12.0 mm (0.48 in), Diameter 6.0 mm (0.24 in)
Iridium
Formula: Ir
MW: 192.22 g/mol
Boiling Pt: 5300 °C (1013 hPa)
Melting Pt: 2450 °C
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS Number: 7439-88-5
EINECS: 231-095-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR