Scandium, arc-cast pellet 15.9 mm (0.63 in) Ø, Diameter 15.9 mm (0.63 in)

Supplier: Thermo Scientific Chemicals

045118-KF
AA45118-KFEA 387.99 USD
AA45118-KF
Scandium, arc-cast pellet 15.9 mm (0.63 in) Ø, Diameter 15.9 mm (0.63 in)
Scandium
Formula: Sc
Boiling Pt: 2730 °C (1013 hPa)
Melting Pt: 1541 °C
Density: 2,989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016323
CAS Number: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR