N-Methylisobutylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
042947-06
42947-06
042947-03
42947-03
AA42947-06EA
76.79
USD
AA42947-06
CAAA42947-03
CAAA42947-06
AA42947-03
N-Methylisobutylamine 98+%
N-Methylisobutylamine
Formula:
C₅H₁₃N Boiling Pt: 78 °C Density: 0,73 Storage Temperature: Ambient |
MDL Number:
MFCD00015043 CAS Number: 625-43-4 EINECS: 210-893-0 UN: 2733 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...