Galena naturally occurring mineral, grains ∼ 1.5-4.8 mm (0.06-0.19 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
042640-14
042640-03
042640-06
42640-03
42640-06
42640-14
AA42640-06EA
39.89
USD
CAAA42640-03
CAAA42640-14
AA42640-06
CAAA42640-06
AA42640-14
AA42640-03
Galena naturally occurring mineral, grains ∼ 1.5-4.8 mm (0.06-0.19 in)
Galena
Galena is used in infrared detectors, photo optic applications, slip property modifier used in friction industry for enhancing heat conduction and regulating friction coefficient, used as a semiconductor. Used in electronic devices and Infrared sensor.
- Hardness: 2.5
- Sp. Gr.: 7.4-7.6
Formula:
PbS MW: 239.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016280 CAS Number: 1314-87-0 EINECS: 215-246-6 UN: 3077 ADR: 9,III Merck Index: 12,05445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...