Galena naturally occurring mineral ∼ 1-2 in
Supplier: Thermo Scientific Chemicals
|
Danger
|
042546-KF
42546-KF
42546-KJ
042546-KJ
AA42546-KJEA
86.09
USD
CAAA42546-KJ
AA42546-KJ
CAAA42546-KF
AA42546-KF
Galena naturally occurring mineral ∼ 1-2 in
Galena
Chief ore of lead
- Sp. Gr. 7.4-7.6
Soluble in strong HNO3, in excess of hot HCl
Formula:
PbS MW: 239.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016280 CAS Number: 1314-87-0 EINECS: 215-246-6 UN: 3077 ADR: 9,III Merck Index: 12,05445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...