Decahydronaphthalene-[D₁₈] (mixture of cis and trans isomers) (99% D)

Danger

Synonyms: Decahydronaphthalene-[²H₁₈] (mixture of cis and trans isomers)

42278-03 42278-03
AA42278-03EA 181.28 USD
CAAA42278-03 AA42278-03
Decahydronaphthalene-[D₁₈] (mixture of cis and trans isomers) (99% D)
Decahydronaphthalene-[D18] (mixture of cis and trans isomers)
Formula: C₁₀D₁₈
MW: 156.11 g/mol
Boiling Pt: 70…71 °C (17 mmHg)
Density: 1.014
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044371
CAS Number: 28788-42-3
UN: 1147
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR