(Triphenylphosphine)gold(I) bromide ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Bromo(triphenylphosphine)gold(I)

41002-03 41002-03
AA41002-03EA 145 USD
CAAA41002-03 AA41002-03
(Triphenylphosphine)gold(I) bromide ≥99.99% (metals basis), Premion®
(Triphenylphosphine)gold (I) bromide
Formula: C₁₈H₁₅AuBrP
Storage Temperature: Ambient
MDL Number: MFCD00049267
CAS Number: 14243-65-3
EINECS: 238-118-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR