Lithium tetrachloroaurate hydrate ≥99.99% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lithium tetrachloroaurate(III) hydrate
40429-03
040429-03
40429-01
40429-01
AA40429-03EA
156.99
USD
AA40429-03
CAAA40429-01
AA40429-01
CAAA40429-03
Lithium tetrachloroaurate hydrate ≥99.99% (metals basis), Premion®
Lithium tetrachloroaurate hydrate
Formula:
AuCl₄Li·H₂O MW: 345.72 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00798533 CAS Number: 3145-91-3 EINECS: 237-190-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...