Zinc (II) cyclohexanebutyrate dihydrate for atomic absorption spectroscopy

Synonyms: Zinc cyclohexanebutyrate dihydrate

39734-06 39734-03 39734-06
AA39734-06EA 206.4 USD
AA39734-06 AA39734-03 CAAA39734-06
Zinc (II) cyclohexanebutyrate dihydrate for atomic absorption spectroscopy
Zinc (II) cyclohexanebutyrate dihydrate
Formula: C₂₀H₃₄O₄Zn·2H₂O
MW: 439.91 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00035537
CAS Number: 38582-18-2
EINECS: 254-017-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR