Tetrachlorobis(tetrahydrofuran)titanium(IV) 98%

Supplier: Thermo Scientific Chemicals
Danger

39594-09 039594-09
AA39594-09EA 196.99 USD
CAAA39594-09 AA39594-09
Tetrachlorobis(tetrahydrofuran)titanium(IV) 98%
Tetrachlorobis(tetrahydrofuran)titanium(IV)
Formula: C₄H₈Cl₄OTi
Melting Pt: 126…128 °C
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077884
CAS Number: 31011-57-1
UN: 2925
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR