Bis(cyclopentadienyl)vanadium dichloride ≥95%

Danger

Synonyms: Vanadocene dichloride

39588-03 39588-03
AA39588-03EA 80.39 USD
AA39588-03 CAAA39588-03
Bis(cyclopentadienyl)vanadium dichloride ≥95%
Bis(cyclopentadienyl)vanadium dichloride
Formula: C₁₀H₁₀Cl₂V
MW: 252.04 g/mol
Melting Pt: 250 °C (dec.)
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00014260
CAS Number: 12083-48-6
EINECS: 235-150-8
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR