Aluminium trifluoromethanesulfonate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum tris(trifluoromethanesulphonate)Aluminum trifluoromethanesulphonateAluminum triflateAluminum trifluoromethanesulfonateAluminum tris(trifluoromethanesulfonate)

038568-09 38568-09
AA38568-09EA 98.39 USD
CAAA38568-09 AA38568-09
Aluminium trifluoromethanesulfonate
Aluminium trifluoromethanesulphonate
Formula: C₃AlF₉O₉S₃
MW: 474.19 g/mol
Melting Pt: ∼ 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00143596
CAS Number: 74974-61-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR