Hydriodic acid 55 - 58%, liquid ACS for use as a catalyst

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen iodideHydroiodic acid

036484-AD 036484-AP 036484-AY
AA36484-AYEA 1085.99 USD
AA36484-AY AA36484-AP AA36484-AD
Hydriodic acid 55 - 58%, liquid ACS for use as a catalyst
Hydriodic acid

Suitable for use in methoxyl determinations.


Used as catalyst for the conversion of primary alcohol to alkyl halide.


Caution: Light sensitive liquid.

Formula: HI
MW: 127.91 g/mol
Boiling Pt: 127 °C (1013 hPa)
Density: 1.701
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,II

Order Now

Specification Test Results

Assay (Unspecified) 55.0 - 58.0%
Residue on Ignition Max. 0.01%
Impurity Content, Chloride and Bromide (as Cl) Max. 0.05%
Impurity Content, Fe Max. 0.001%
Impurity Content, I₂ Max. 0.75%
Impurity Content, PO₄ Max. 0.001%
Impurity Content, SO₄ Max. 0.005%
Trace Metals (as Pb) Max. 0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR