(S)-(+)-2-Heptanol ≥98%

Warning

33795-06 33795-03 33795-MD 33795-03 33795-MD
AA33795-06EA 223 USD
AA33795-06 AA33795-MD CAAA33795-MD AA33795-03 CAAA33795-03
(S)-(+)-2-Heptanol ≥98%
(S)-(+)-2-Heptanol
Formula: C₇H₁₆O
MW: 116.2 g/mol
Boiling Pt: 160…162 °C
Density: 0.815
Flash Pt: 59 °C (138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065003
CAS Number: 6033-23-4
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR