Lutetium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
14566-03
014566-03
AA14566-03EA
165.34
USD
AA14566-03
CAAA14566-03
Lutetium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Lutetium (III) sulphate octahydrate
Formula:
Lu₂O₁₂S₃·8H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00011499 CAS Number: 13473-77-3 EINECS: 239-074-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...