Dipraseodymium tricarbonate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Alfa Aesar

Synonyms: Praseodymium(III) carbonate octahydrate

13182-22 13182-06 13182-06 13182-14 13182-14 13182-22
AA13182-06EA 30.77 USD
CAAA13182-22 CAAA13182-06 AA13182-06 CAAA13182-14 AA13182-14 AA13182-22
Dipraseodymium tricarbonate octahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dipraseodymium tricarbonate octahydrate
Formula: C₃O₉Pr₂·8H₂O
Storage Temperature: Ambient
MDL Number: MFCD00211307
CAS Number: 14948-62-0
EINECS: 227-578-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR