Hexachloroiridic(IV) acid hydrate (38 - 42% Ir) ≥99% (metals basis) Ir 38-42%

Danger

Synonyms: Dihydrogen hexachloroiridate(IV) hydrateHexachloroiridium(IV) acid hydrateHydrogen hexachloroiridate(IV) hydrateHydrogen iridium hexachloride

11031-06 11031-03 11031-14 11031-03 11031-14 11031-06
AA11031-06EA 1021.46 USD
AA11031-06 AA11031-03 AA11031-14 CAAA11031-06 CAAA11031-03 CAAA11031-14
Hexachloroiridic(IV) acid hydrate (38 - 42% Ir) ≥99% (metals basis) Ir 38-42%
Hexachloroiridic (IV) acid

Soluble in alcohol and water

Formula: H₂Cl₆Ir·H₂O
MW: 406.93 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00011328
CAS Number: 57876-28-5
EINECS: 241-012-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR