Flammable Storage Laboratory Refrigerator, Manual Defrost, Nor-Lake® Scientific
NSFR241WMW0M
89368-152EA 5258 USD
89368-152
Flammable Storage Laboratory Refrigerator, Manual Defrost, Nor-Lake® Scientific
Designed for the storage of flammables in approved containers for Class I, Division 2 Storage of Flammable Liquids, per NFPA Standards 70, 99, 45, 497A, and UL471
Refrigerators feature mechanical cold controller and internal chamber thermometer with adjustable temperature control range from 2 to 10°C (36 to 50°F). The refrigerator is factory preset at 4°C (39°F). The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner and interior door panel are smooth aluminum. Cabinet and solid door are foamed-in-place with CFC-free, high-density polyurethane foam insulation. The heavy-duty solid door features spring-loaded hinges, full-length pull door handle, magnetic door gasket, and centered key door lock (2 keys included). Additional standard features include four casters (2 locking), three coated wire shelves, air cooled condensing unit, top-mounted refrigeration system, and cold wall evaporator. Manual defrost.
Certifications: UL and C-UL listed.
Ordering information: Refrigerator is supplied with 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).