Moticam® 10+ Microscope Camera, Motic Instruments
Supplier: Motic
1100600100871
75784-570EA 1773.26 USD
75784-570
Moticam® 10+ Microscope Camera, Motic Instruments
The Moticam® 10+ is Motic's highest resolution microscope camera. Designed to produce publication quality images, the Moticam® 10+ offers a 10 megapixel live and capture resolution for easy conversion of 300dpi documentation files.
  • 10 megapixel resolution results in print publication worthy images
  • Motic's Rapid View Algorithm enables high framerate live viewing while enabling full resolution image capture
  • Convert captured images to 300dpi documentation files easily
  • USB 3 connection ensures the fastest data transfer speeds
  • Motic Images Plus software is incuded for viewing, measurement, annotation and reporting of images
  • Comes with multiple accessories to get you started right away, including calibration slide, macro viewing tube eyepiece adapters, software, cables and more
Motic's rapid view algorithm enables the user to display a faster frame rate at full screen, while maintaining the capability to capture still images at maximum resolution
The Moticam® 10+ is Motic's flagship digital maicroscope camera. Boasting an amazing 10 megapixel resolution, the Moticam® 10+ is the perfect choice for those in need of publication quality microscope photos and those who want to view their samples at the highest level of detail possible.
The Moticam® 10+'s flexbility and ease of use is enhanced by Motic's rapid view algorithm, which enables the user to view samples at a very high frame rate, without sacrificing image capture quality. At the same time, a USB 3 connection enables lightining fast data transfer speeds.
The lastest version of Motic Images Plus software is included, giving users the ability to not only view their images, but also measure, annotate, report and adjust them as well. No license fees or upgrade fees necessary. Just register your Moticam® online and receive free updates as they are made available.
Recently upgraded to USB3 technology, the Moticam® 10+ data transfer speeds are faster than ever. The Moticam® 10+ is perfect for professionals desiring flexibility and the highest quality microscope images.
Certifications: UL Listed.
Ordering information: Camera is supplied with all accessories listed above. Contact your VWR representative to learn more about this and other Motic solutions. All Motic products come with a 1 year electrical and 5 year mechanical warranty.