(S)-(+)-2-Octyl isocyanate 95%

Supplier: Thermo Scientific Chemicals

Warning

L20245-03 L20245-03
AAAL20245-03EA 132 USD
CAAAAL20245-03 AAAL20245-03
(S)-(+)-2-Octyl isocyanate 95%
(S)-(+)-2-Octyl isocyanate
Formula: C₉H₁₇NO
Boiling Pt: 68 °C/5 mm
Density: 0.826
Storage Temperature: Ambient
MDL Number: MFCD05664089
CAS Number: 745783-87-3
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR