(R)-(-)-2-Octyl isocyanate 90+%

Warning

L20244-03 L20244-03
AAAL20244-03EA 138.75 USD
CAAAAL20244-03 AAAL20244-03
(R)-(-)-2-Octyl isocyanate 90+%
(R)-(-)-2-Octyl isocyanate
Formula: C₉H₁₇NO
Boiling Pt: 68 °C/5 mm
Density: 0.826
Storage Temperature: Ambient
MDL Number: MFCD05664088
CAS Number: 745783-86-2
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR