(R)-(-)-2-Nonyl isocyanate 97%

Warning

L20236-03 L20236-03
AAAL20236-03EA 127.83 USD
CAAAAL20236-03 AAAL20236-03
(R)-(-)-2-Nonyl isocyanate 97%
(R)-(-)-2-Nonyl isocyanate
Formula: C₁₀H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD05664081
EINECS: 000-000-0
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR