(R)-(-)-2-Hexyl isocyanate 95%

Danger

L20234-03 L20234-03
AAAL20234-03EA 119.88 USD
AAAL20234-03 CAAAAL20234-03
(R)-(-)-2-Hexyl isocyanate 95%
(R)-(-)-2-Hexyl isocyanate
Formula: C₇H₁₃NO
Flash Pt: 47 °C(117 °F)
Storage Temperature: Ambient
MDL Number: MFCD05664079
UN: 3080
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR