(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate 95%

Supplier: Thermo Scientific Chemicals

Warning

L20231-03 L20231-03
AAAL20231-03EA 108 USD
CAAAAL20231-03 AAAL20231-03
(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate 95%
(S)-(-)-1-(4-Fluorophenyl)ethyl isocyanate
Formula: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD05664076
CAS Number: 745783-74-8
EINECS: 000-000-0
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR