(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate tech. 90%, Technical Grade

Warning

L20227-03 L20227-03
AAAL20227-03EA 91.02 USD
CAAAAL20227-03 AAAL20227-03
(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate tech. 90%, Technical Grade
(S)-(-)-1-(4-Chlorophenyl)ethyl isocyanate
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD05664072
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR