4'-tert-Butyl-2-chloroacetophenone 97%

Warning

Synonyms: 4-(tert-Butyl)phenacyl chloride

L20156-06 L20156-03 L20156-06 L20156-03
AAAL20156-06EA 243.09 USD
AAAL20156-06 CAAAAL20156-06 AAAL20156-03 CAAAAL20156-03
4'-tert-Butyl-2-chloroacetophenone 97%
4'-tert-Butyl-2-chloroacetophenone
Formula: C₉H₉ClO
MW: 210.71
Boiling Pt: 142…146 °C/1 mm
Melting Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00171621
CAS Number: 21886-62-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR