(S)-(+)-1-(3-Methoxyphenyl)ethyl isothiocyanate 98%

Danger

L20069-03 L20069-03
AAAL20069-03EA 91.16 USD
CAAAAL20069-03 AAAL20069-03
(S)-(+)-1-(3-Methoxyphenyl)ethyl isothiocyanate 98%
(S)-(+)-1-(3-Methoxyphenyl)ethyl isothiocyanate
Formula: C₁₀H₁₁NOS
Density: 1.120
Storage Temperature: Ambient
MDL Number: MFCD05664056
CAS Number: 737000-99-6
EINECS: 000-000-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR