(S)-(+)-2-Heptyl isothiocyanate 98%
|
Danger
|
L20065-03
L20065-03
AAAL20065-03EA
98.17
USD
AAAL20065-03
CAAAAL20065-03
(S)-(+)-2-Heptyl isothiocyanate 98%
(S)-(+)-2-Heptyl isothiocyanate
Formula:
C₈H₁₅NS Density: 0.910 Storage Temperature: Ambient |
EINECS:
000-000-0 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...