(R)-(-)-2-Heptyl isothiocyanate 98%
|
Danger
|
L20064-03
L20064-03
AAAL20064-03EA
98.17
USD
AAAL20064-03
CAAAAL20064-03
(R)-(-)-2-Heptyl isothiocyanate 98%
(R)-(-)-2-Heptyl isothiocyanate
Formula:
C₈H₁₅NS Density: 0.910 Storage Temperature: Ambient |
EINECS:
000-000-0 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...