2-Methoxy-6-(methylamino)pyridine 98%

Warning

L20048-14 L20048-14 L20048-06 L20048-06
AAAL20048-06EA 52.99 USD
AAAL20048-06 CAAAAL20048-14 AAAL20048-14 CAAAAL20048-06
2-Methoxy-6-(methylamino)pyridine 98%
2-Methoxy-6-(methylamino)pyridine
Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191557
CAS Number: 88569-83-9
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR