2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine 95%

Warning

L20009-MC L20009-MC L20009-ME L20009-ME
AAAL20009-MCEA 106.14 USD
CAAAAL20009-MC AAAL20009-MC CAAAAL20009-ME AAAL20009-ME
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine 95%
2-Amino-3,6-difluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₅N₂
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD03840200
CAS Number: 675602-89-8
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR