(S)-(+)-2-Octyl isothiocyanate 97%

Danger

L19985-03 L19985-03
AAAL19985-03EA 91.68 USD
CAAAAL19985-03 AAAL19985-03
(S)-(+)-2-Octyl isothiocyanate 97%
(S)-(+)-2-Octyl isothiocyanate
Formula: C₉H₁₇NS
MW: 171.31 g/mol
Boiling Pt: 95…96 °C/4 mm
Density: 0.907
Storage Temperature: Ambient
MDL Number: MFCD04039337
EINECS: 000-000-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR