(R)-(-)-2-Octyl isothiocyanate 98%

Danger

L19984-03 L19984-03
AAAL19984-03EA 117.66 USD
CAAAAL19984-03 AAAL19984-03
(R)-(-)-2-Octyl isothiocyanate 98%
(R)-(-)-2-Octyl isothiocyanate
Formula: C₉H₁₇NS
Boiling Pt: 95…96 °C/4 mm
Density: 0.907
Storage Temperature: Ambient
MDL Number: MFCD04039336
CAS Number: 21663-52-5
EINECS: 000-000-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR