(S)-(+)-2-Nonyl isothiocyanate 97%
|
Danger
|
L19983-03
L19983-03
AAAL19983-03EA
91.68
USD
CAAAAL19983-03
AAAL19983-03
(S)-(+)-2-Nonyl isothiocyanate 97%
(S)-(+)-2-Nonyl isothiocyanate
Formula:
C₁₀H₁₉NS Density: 0.904 Storage Temperature: Ambient |
EINECS:
000-000-0 UN: 2922 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...