3-Cyano-4-fluorobenzenesulfonyl chloride 98%

Danger

L19885-03 L19885-06 L19885-MD L19885-03 L19885-06 L19885-MD
AAAL19885-03EA 105.89 USD
CAAAAL19885-03 CAAAAL19885-06 AAAL19885-03 AAAL19885-06 CAAAAL19885-MD AAAL19885-MD
3-Cyano-4-fluorobenzenesulfonyl chloride 98%
3-Cyano-4-fluorobenzenesulphonyl chloride
Formula: C₇H₃ClFNO₂S
MW: 219.62 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD03093760
CAS Number: 351003-23-1
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR