3-(Boc-amino)-3-(3-fluorophenyl)propionic acid 99%

Synonyms: 3-[(tert-Butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid3-((tert-Butoxycarbonyl)amino)-3-(3-fluorophenyl)propanoic acid

L19805-03 L19805-MD L19805-03 L19805-MD
AAAL19805-MDEA 51.49 USD
CAAAAL19805-MD AAAL19805-MD CAAAAL19805-03 AAAL19805-03
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid 99%
3-(Boc-amino)-3-(3-fluorophenyl)propionic acid
Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD02090701
CAS Number: 284493-59-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR