3-(Boc-amino)-3-(2-fluorophenyl)propionic acid 98+%

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(2-fluorophenyl)propanoic acid

L19804-MD L19804-MD L19804-03 L19804-03
AAAL19804-MDEA 46.45 USD
AAAL19804-MD CAAAAL19804-MD CAAAAL19804-03 AAAL19804-03
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid 98+%
3-(Boc-amino)-3-(2-fluorophenyl)propionic acid
Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD02090698
CAS Number: 284493-56-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR