3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%

Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

L19803-03 L19803-03 L19803-MD L19803-MD
AAAL19803-03EA 166.53 USD
CAAAAL19803-MD AAAL19803-03 AAAL19803-MD CAAAAL19803-03
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid 97%
3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
Formula: C₁₄H₁₈ClNO₄
MW: 299.75 g/mol
Melting Pt: 179…180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02090708
CAS Number: 284493-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR