3-Amino-3-(3-methoxyphenyl)propionic acid 97%

Synonyms: 3-Amino-3-(3-methoxyphenyl)propanoic acid

L19800-MD L19800-MD L19800-03 L19800-03
AAAL19800-03EA 89.59 USD
AAAL19800-03 CAAAAL19800-03 AAAL19800-MD CAAAAL19800-MD
3-Amino-3-(3-methoxyphenyl)propionic acid 97%
3-Amino-3-(3-methoxyphenyl)propionic acid
Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
Melting Pt: 208 °C (dec.)
Storage Temperature: Ambient
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR