3-Amino-3-(4-chlorophenyl)propionic acid 97%

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid3-Amino-3-(p-chlorophenyl)propionic acid

L19798-MD L19798-06 L19798-MD L19798-06 L19798-03 L19798-03
AAAL19798-MDEA 29.46 USD
CAAAAL19798-03 AAAL19798-MD CAAAAL19798-06 CAAAAL19798-MD AAAL19798-06 AAAL19798-03
3-Amino-3-(4-chlorophenyl)propionic acid 97%
3-Amino-3-(4-chlorophenyl)propionic acid
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
Melting Pt: 214…216 °C (dec.)
Storage Temperature: Ambient
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR