1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 8-Amino-1,4-dioxaspiro[4.5]decane-8-carboxylic Acid

L19795-MD L19795-MD L19795-03 L19795-03
AAAL19795-MDEA 63.49 USD
AAAL19795-MD CAAAAL19795-MD AAAL19795-03 CAAAAL19795-03
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal 98%
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal
Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
Melting Pt: 301…304 °C
Storage Temperature: Ambient
MDL Number: MFCD03790906
CAS Number: 54621-18-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR